Anti-MSMB Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: MSMB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77743-531
- 77860-043
- Antigen Symbol:
- MSMB
- MSMB
- Antigen Synonyms:
- Beta microseminoprotein [Precursor]|Beta-microseminoprotein|HPC 13|HPC13|IGBF|Immunoglobulin binding factor|Immunoglobulin-binding factor|microseminoprotein beta|MSMB_HUMAN|MSP|MSPB|PN 44|PN44|Prostate secreted seminal plasma protein|Prostate secretory protein of 94 amino acids|Prostate secretory protein PSP94|PRPS|PRSP|PSP 57|PSP|PSP-94|PSP57|PSP94|Seminal plasma beta inhibin|Seminal plasma beta-inhibin
- Beta microseminoprotein [Precursor]|Beta-microseminoprotein|HPC 13|HPC13|IGBF|Immunoglobulin binding factor|Immunoglobulin-binding factor|microseminoprotein beta|MSMB_HUMAN|MSP|MSPB|PN 44|PN44|Prostate secreted seminal plasma protein|Prostate secretory protein of 94 amino acids|Prostate secretory protein PSP94|PRPS|PRSP|PSP 57|PSP|PSP-94|PSP57|PSP94|Seminal plasma beta inhibin|Seminal plasma beta-inhibin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08118
- P08118
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Beta-microseminoprotein (27-107AA)
- Recombinant Human Beta-microseminoprotein (27-107AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together