Anti-IFNA14 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFNA14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-671
- 77857-875
- Antigen Symbol:
- IFNA14
- IFNA14
- Antigen Synonyms:
- IFN-alpha-14|IFN14_HUMAN|IFNA14|Interferon alpha 14|Interferon alpha H|Interferon alpha-14|Interferon alpha-H|Interferon lambda-2-H|Interferon lambda2H |LeIF H|LeIFH|MGC125756|MGC125757
- IFN-alpha-14| IFN14_HUMAN| IFNA14| Interferon alpha 14| Interferon alpha H| Interferon alpha-14| Interferon alpha-H| Interferon lambda-2-H| Interferon lambda2H | LeIF H| LeIFH| MGC125756| MGC125757
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01570
- P01570
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon alpha-14 protein (24-189AA)
- Recombinant Human Interferon alpha-14 protein (24-189AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together