Anti-ARAF Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARAF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-629
- 77745-630
- Antigen Symbol:
- ARAF
- ARAF
- Antigen Synonyms:
- A raf 1|A Raf proto oncogene serine/threonine protein kinase|ARAF 1|ARaf proto oncogene serine/threonine protein kinase|ARAF_HUMAN|ARAF1|Oncogene Araf1|Oncogene PKS2|PKS 2|PKS|PKS2|Proto oncogene Pks|Proto-oncogene A-Raf|Proto-oncogene A-Raf-1|Proto-oncogene Pks|RAFA 1|RAFA1|Ras binding protein DA Raf|Serine/threonine-protein kinase A-Raf|v raf murine sarcoma 3611 viral oncogene homolog|v raf murine sarcoma 3611 viral oncogene homolog 1|v raf oncogene homolog 1 (murine sarcoma 3611 virus)
- A raf 1|A Raf proto oncogene serine/threonine protein kinase|ARAF 1|ARaf proto oncogene serine/threonine protein kinase|ARAF_HUMAN|ARAF1|Oncogene Araf1|Oncogene PKS2|PKS 2|PKS|PKS2|Proto oncogene Pks|Proto-oncogene A-Raf|Proto-oncogene A-Raf-1|Proto-oncogene Pks|RAFA 1|RAFA1|Ras binding protein DA Raf|Serine/threonine-protein kinase A-Raf|v raf murine sarcoma 3611 viral oncogene homolog|v raf murine sarcoma 3611 viral oncogene homolog 1|v raf oncogene homolog 1 (murine sarcoma 3611 virus)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10398
- P10398
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Serine/threonine-protein kinase A-Raf protein (160-310AA)
- Serine/threonine-protein kinase A-Raf protein (160-310AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together