Anti-GARS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: GARS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77737-981
- 77855-804
- Antigen Symbol:
- GARS
- GARS
- Antigen Synonyms:
- GARS1|GARSGlycine--tRNA ligase|EC 6.1.1.14|Diadenosine tetraphosphate synthetase|Ap4A synthetase|EC 2.7.7.-|Glycyl-tRNA synthetase|GlyRS|Glycyl-tRNA synthetase 1
- GARS1|GARSGlycine--tRNA ligase|EC 6.1.1.14|Diadenosine tetraphosphate synthetase|Ap4A synthetase|EC 2.7.7.-|Glycyl-tRNA synthetase|GlyRS|Glycyl-tRNA synthetase 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41250
- P41250
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glycine--tRNA ligase protein (1-389AA)
- Recombinant Human Glycine--tRNA ligase protein (1-389AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together