Anti-MOCS2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IP: 1:200 to 1:2000, IF: 1:50 to 1:200
Type:
Antigen: MOCS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-292
- 77859-876
- Antigen Symbol:
- MOCS2
- MOCS2
- Antigen Synonyms:
- MCBPE|MOCO1|Molybdopterin synthase catalytic subunit|EC 2.8.1.12|MOCO1-B|Molybdenum cofactor synthesis protein 2 large subunit|Molybdenum cofactor synthesis protein 2B|MOCS2B|Molybdopterin-synthase large subunit|MPT synthase large subunit
- MCBPE|MOCO1|Molybdopterin synthase catalytic subunit|EC 2.8.1.12|MOCO1-B|Molybdenum cofactor synthesis protein 2 large subunit|Molybdenum cofactor synthesis protein 2B|MOCS2B|Molybdopterin-synthase large subunit|MPT synthase large subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O96007
- O96007
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Molybdopterin synthase catalytic subunit protein (1-188AA)
- Recombinant human Molybdopterin synthase catalytic subunit protein (1-188AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together