Anti-EXO1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: EXO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77854-995
- 77736-273
- Antigen Symbol:
- EXO1
- EXO1
- Antigen Synonyms:
- exo1| EXO1_HUMAN| ExoI| Exonuclease 1| Exonuclease I| Exonuclease1| HEX1| hExo I| hExo1| hExoI| Rad2 nuclease family member homolog of S. cerevisiae exonuclease 1
- exo1|EXO1_HUMAN|ExoI|Exonuclease 1|Exonuclease I|Exonuclease1|HEX1|hExo I|hExo1|hExoI|Rad2 nuclease family member homolog of S. cerevisiae exonuclease 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UQ84
- Q9UQ84
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Exonuclease 1 protein (301-600AA)
- Recombinant Human Exonuclease 1 protein (301-600AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together