Anti-KCNN1 Rabbit Polyclonal Antibody
77741-523
New Product
- Antigen Symbol:KCNN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q92952
- Antigen Synonyms:KCNN1|SKSmall conductance calcium-activated potassium channel protein 1|SK1|SKCa 1|SKCa1|KCa2.1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:KCa2.1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: KCNN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat