Anti-GNAT2 Rabbit Polyclonal Antibody
Catalog # 77737-529
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GNAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P19087
- Antigen Synonyms:ACHM4|Cone type transducin alpha subunit|GNAT 2|GNAT C|Gnat2|GNAT2_HUMAN|GNATC|Guanine nucleotide binding protein (G protein) alpha transducing activity polypeptide 2|Guanine nucleotide binding protein G t subunit alpha 2|Guanine nucleotide-binding protein G(t) subunit alpha-2|Transducin alpha 2|Transducin alpha-2 chain|Transducin alpha2|Transducin cone specific alpha polypeptide
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:ACHM4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: GNAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse