Anti-TRPM2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: TRPM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77736-115
- 77854-890
- Antigen Symbol:
- TRPM2
- TRPM2
- Antigen Synonyms:
- EREG 1|EREG1|Estrogen responsive element associated gene 1|Estrogen responsive element associated gene 1 protein|Estrogen-responsive element-associated gene 1 protein|KNP 3|KNP3|Long transient receptor potential channel 2|LTRPC 2|LTrpC-2|LTrpC2|MGC133383|NUDT9H|NUDT9L1|putative Ca2+ channel protein|Transient receptor potential cation channel subfamily M member 2|Transient receptor potential channel 7|TRPC 7|TrpC7|TRPM 2|Trpm2|TRPM2_HUMAN
- EREG 1| EREG1| Estrogen responsive element associated gene 1| Estrogen responsive element associated gene 1 protein| Estrogen-responsive element-associated gene 1 protein| KNP 3| KNP3| Long transient receptor potential channel 2| LTRPC 2| LTrpC-2| LTrpC2| MGC133383| NUDT9H| NUDT9L1| putative Ca2+ channel protein| Transient receptor potential cation channel subfamily M member 2| Transient receptor potential channel 7| TRPC 7| TrpC7| TRPM 2| Trpm2| TRPM2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94759
- O94759
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transient receptor potential cation channel subfamily M member 2 protein (60-119AA)
- Recombinant Human Transient receptor potential cation channel subfamily M member 2 protein (60-119AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together