Anti-KEAP1 Rabbit Polyclonal Antibody
Catalog # 77741-551
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KEAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14145
- Antigen Synonyms:Cytosolic inhibitor of Nrf2|INrf 2|INrf2|Keap 1|KEAP1_HUMAN|Kelch like ECH associated protein 1|Kelch like family member 19|Kelch like protein 19|Kelch-like ECH-associated protein 1|Kelch-like protein 19|KIAA0132|KLHL 19|KLHL19|MGC10630|MGC1114|MGC20887|MGC4407|MGC9454
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MGC9454
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KEAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat