Anti-NBR1 Rabbit Polyclonal Antibody
77743-968
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Symbol:NBR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14596
- Antigen Synonyms:1A13B|Neighbor of BRCA1 gene 1 protein|Membrane component|Migration inducing protein 19|NBR 1|1A1 3B|M17S2|Neighbor of BRCA1 gene 1|surface marker 2 (ovarian carcinoma antigen CA125)|Membrane component chromosome 17 surface marker 2|MIG19|NBR1_HUMAN|Ovarian carcinoma antigen CA125|Protein 1A1-3B|Cell migration-inducing gene 19 protein|KIAA0049|MIG 19|Next to BRCA1 gene 1 protein|autophagy cargo receptor|B box protein|chromosome 17
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Protein 1A1-3B
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat