Anti-HBZ Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: HBZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-775
- 77739-484
- Antigen Symbol:
- HBZ
- Antigen Synonyms:
- HBAZ|HBAZ_HUMAN|HBZ 2|HBZ|HBZ2|Hemoglobin subunit zeta|Hemoglobin zeta|Hemoglobin zeta chain|Zeta globin|Zeta-globin
- HBAZ|HBAZ_HUMAN|HBZ 2|HBZ|HBZ2|Hemoglobin subunit zeta|Hemoglobin zeta|Hemoglobin zeta chain|Zeta globin|Zeta-globin
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P02008
- P02008
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Hemoglobin subunit zeta protein (1-142AA)
- Recombinant Human Hemoglobin subunit zeta protein (1-142AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together