Anti-HELLS Rabbit Polyclonal Antibody
Catalog # 77739-619
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HELLS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NRZ9
- Antigen Synonyms:FLJ10339|Helicase lymphoid specific|HELLS_HUMAN|LSH|Lymphoid specific helicase|Lymphoid-specific helicase|Nbla10143|PASG|Proliferation associated SNF2 like protein|Proliferation-associated SNF2-like protein|SWI/SNF2 related matrix associated actin dependent regulator of chromatin subfamily A member 6|SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HELLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat