Anti-GNG11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNG11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-590
- 77856-219
- Antigen Symbol:
- GNG11
- GNG11
- Antigen Synonyms:
- G protein gamma 11 subunit|GBG11_HUMAN|Gng11|GNGT11|Guanine nucleotide binding protein G|Guanine nucleotide binding protein gamma 11|Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11|IG|Osubunit gamma 11 precursor|SG
- G protein gamma 11 subunit|GBG11_HUMAN|Gng11|GNGT11|Guanine nucleotide binding protein G|Guanine nucleotide binding protein gamma 11|Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11|IG|Osubunit gamma 11 precursor|SG
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61952
- P61952
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11 protein (1-70AA)
- Recombinant Human Guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-11 protein (1-70AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together