Anti-HOXB3 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:500-1:1000
Type:
Antigen: HOXB3
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-577
- 77740-295
- Antigen Symbol:
- HOXB3
- HOXB3
- Antigen Synonyms:
- Homeo box 2G|Homeo box B3|Homeobox B3|Homeobox protein Hox-2.7|Homeobox protein Hox-2G|Homeobox protein Hox-B3|Homeobox protein Hox 2.7|HOX 2G|HOX2|Hox2.7|HOX2G|HXB3_HUMAN
- Homeo box 2G| Homeo box B3| Homeobox B3| Homeobox protein Hox-2.7| Homeobox protein Hox-2G| Homeobox protein Hox-B3| Homeobox protein Hox 2.7| HOX 2G| HOX2| Hox2.7| HOX2G| HXB3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14651
- P14651
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-B3 protein (315-423AA)
- Recombinant Human Homeobox protein Hox-B3 protein (315-423AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together