Anti-HSD17B1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HSD17B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-416
- 77857-680
- Antigen Symbol:
- HSD17B1
- HSD17B1
- Antigen Synonyms:
- 17 beta HSD 1|17 beta hydroxysteroid dehydrogenase type 1|17-beta-HSD 1|17-beta-hydroxysteroid dehydrogenase type 1|20 alpha-hydroxysteroid dehydrogenase|20-alpha-HSD|DHB1_HUMAN|E17KSR|E2DH|EDH17B1|EDH17B2|EDHB17|Estradiol 17 beta dehydrogenase 1|Estradiol 17-beta-dehydrogenase 1|HSD17|HSD17B1|Hydroxysteroid (17 beta) dehydrogenase 1|MGC13814|Placental 17 beta hydroxysteroid dehydrogenase|Placental 17-beta-hydroxysteroid dehydrogenase|SDR28C1|Short chain dehydrogenase/reductase family 28CE,member 1
- 17 beta HSD 1| 17 beta hydroxysteroid dehydrogenase type 1| 17-beta-HSD 1| 17-beta-hydroxysteroid dehydrogenase type 1| 20 alpha-hydroxysteroid dehydrogenase| 20-alpha-HSD| DHB1_HUMAN| E17KSR| E2DH| EDH17B1| EDH17B2| EDHB17| Estradiol 17 beta dehydrogenase 1| Estradiol 17-beta-dehydrogenase 1| HSD17| HSD17B1| Hydroxysteroid (17 beta) dehydrogenase 1| MGC13814| Placental 17 beta hydroxysteroid dehydrogenase| Placental 17-beta-hydroxysteroid dehydrogenase| SDR28C1| Short chain dehydrogenase/reductase family 28CE,member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14061
- P14061
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Estradiol 17-beta-dehydrogenase 1 protein (268-328AA)
- Recombinant Human Estradiol 17-beta-dehydrogenase 1 protein (268-328AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together