Anti-DGKB Rabbit Polyclonal Antibody
Catalog # 77735-837
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DGKB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y6T7
- Antigen Synonyms:90 kDa diacylglycerol kinase|DAG kinase beta|DAGK2|DGK|DGK B|DGK beta|DGK-beta|DGKB_HUMAN|Diacylglycerol kinase beta (90kD)|Diacylglycerol kinase beta 90kDa|Diacylglycerol kinase beta|Diglyceride kinase beta|KIAA0718
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:90 kDa diacylglycerol kinase
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: DGKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat