Anti-KRT6A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT6A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-754
- 77741-891
- Antigen Symbol:
- KRT6A
- KRT6A
- Antigen Synonyms:
- CK-6A|CK-6D|CK6A|CK6C|CK6D|Cytokeratin-6A|Cytokeratin-6D|K2C6A_HUMAN|K6A|K6C|K6D|keratin 6A|Keratin|type II cytoskeletal 6A|Keratin-6A|Krt6a|KRT6C|KRT6D|Type-II keratin Kb6
- CK-6A|CK-6D|CK6A|CK6C|CK6D|Cytokeratin-6A|Cytokeratin-6D|K2C6A_HUMAN|K6A|K6C|K6D|keratin 6A|Keratin|type II cytoskeletal 6A|Keratin-6A|Krt6a|KRT6C|KRT6D|Type-II keratin Kb6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02538
- P02538
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Keratin, type II cytoskeletal 6A protein (2-564AA)
- Recombinant Human Keratin, type II cytoskeletal 6A protein (2-564AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together