Anti-NPBWR2 Rabbit Recombinant polyclonal Antibody
New Product
Recommended dilution: IF:1:50-1:200
Type:
Antigen: NPBWR2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77744-754
- 77744-753
- Antigen Symbol:
- NPBWR2
- NPBWR2
- Antigen Synonyms:
- GPR8| Neuropeptides B/W receptor type 2| G-protein coupled receptor 8
- GPR8| Neuropeptides B/W receptor type 2| G-protein coupled receptor 8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48146
- P48146
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Neuropeptides B/W receptor type 2 protein (1-45AA)
- Neuropeptides B/W receptor type 2 protein (1-45AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together