Anti-MAP1B Rabbit Polyclonal Antibody
Catalog # 77742-623
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAP1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P46821
- Antigen Synonyms:FUTSCH|LC1|MAP-1B|MAP1 light chain LC1|Map1b|MAP1B_HUMAN|MAP5|Microtubule associated protein 1B|Mtap1b|Mtap5
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Mtap5
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MAP1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat