Anti-MOCS3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: MOCS3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-297
- 77859-881
- Antigen Symbol:
- MOCS3
- MOCS3
- Antigen Synonyms:
- Adenylyltransferase dJ914P20.3|MGC9252|MOCS 3|MOCS3_HUMAN|Molybdenum cofactor synthesis 3|Molybdenum cofactor synthesis protein 3|Molybdopterin synthase sulfurylase|Molybdopterin-synthase sulfurtransferase|MPT synthase sulfurylase|Sulfur carrier protein MOCS2A adenylyltransferase|Sulfur carrier protein MOCS2A sulfurtransferase|Sulfurtransferase UBA 4|UBA4 ubiquitin activating enzyme E1 homolog|UBA4
- Adenylyltransferase dJ914P20.3|MGC9252|MOCS 3|MOCS3_HUMAN|Molybdenum cofactor synthesis 3|Molybdenum cofactor synthesis protein 3|Molybdopterin synthase sulfurylase|Molybdopterin-synthase sulfurtransferase|MPT synthase sulfurylase|Sulfur carrier protein MOCS2A adenylyltransferase|Sulfur carrier protein MOCS2A sulfurtransferase|Sulfurtransferase UBA 4|UBA4 ubiquitin activating enzyme E1 homolog|UBA4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95396
- O95396
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Adenylyltransferase and sulfurtransferase MOCS3 protein (271-460AA)
- Recombinant human Adenylyltransferase and sulfurtransferase MOCS3 protein (271-460AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together