Anti-LSM8 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: LSM8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-433
- 77859-203
- Antigen Symbol:
- LSM8
- LSM8
- Antigen Synonyms:
- LSM8U6 snRNA-associated Sm-like protein LSm8
- LSM8U6 snRNA-associated Sm-like protein LSm8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95777
- O95777
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human U6 snRNA-associated Sm-like protein LSm8 protein (2-96AA)
- Recombinant Human U6 snRNA-associated Sm-like protein LSm8 protein (2-96AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together