Anti-NAA16 Rabbit Polyclonal Antibody
Catalog # 77743-925
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NAA16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6N069
- Antigen Synonyms:NARG1-like protein|NMDA receptor-regulated 1-like protein|NatA auxiliary subunit|NAT2|NARG1L|N-alpha-acetyltransferase 16
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:NARG1L
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NAA16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat