Anti-MYO1C Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MYO1C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-221
- 77743-775
- Antigen Symbol:
- MYO1C
- MYO1C
- Antigen Synonyms:
- MMI beta|MMI-beta|MMIb|MYO1C_HUMAN|Myosin I beta|Myosin Ic|Myosin-Ic|MYR2|NMI|Nuclear myosin I
- MMI beta|MMI-beta|MMIb|MYO1C_HUMAN|Myosin I beta|Myosin Ic|Myosin-Ic|MYR2|NMI|Nuclear myosin I
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00159
- O00159
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Unconventional myosin-Ic protein (804-1063AA)
- Recombinant human Unconventional myosin-Ic protein (804-1063AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together