Anti-AGFG1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: AGFG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77744-567
- 77744-566
- Antigen Symbol:
- AGFG1
- AGFG1
- Antigen Synonyms:
- AGFG1|AGFG1_HUMAN|Arf-GAP domain and FG repeats-containing protein 1|ArfGAP with FG repeats 1|AU045498|C130049H11Rik|C85612|D730048C23Rik|DKFZp686I15205|HIV 1 Rev binding protein|HIV-1 Rev-binding protein|HRB|MGC116375|MGC116938|MGC116940|Nucleoporin like protein RIP|Nucleoporin-like protein RIP|RAB|Rev interacting protein|Rev-interacting protein|Rev/Rex activation domain-binding protein|RIP
- AGFG1|AGFG1_HUMAN|Arf-GAP domain and FG repeats-containing protein 1|ArfGAP with FG repeats 1|AU045498|C130049H11Rik|C85612|D730048C23Rik|DKFZp686I15205|HIV 1 Rev binding protein|HIV-1 Rev-binding protein|HRB|MGC116375|MGC116938|MGC116940|Nucleoporin like protein RIP|Nucleoporin-like protein RIP|RAB|Rev interacting protein|Rev-interacting protein|Rev/Rex activation domain-binding protein|RIP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52594
- P52594
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of AGFG1
- Fusion protein of AGFG1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together