Anti-HARS2 Rabbit Recombinant polyclonal Antibody (Biotinylated)
New Product
0
Type:
Antigen: HARS2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77739-422
- 77856-722
- Antigen Symbol:
- HARS2
- HARS2
- Antigen Synonyms:
- 4631412B19Rik| AI593507| HARS related| HARSL| HARSR| HisRS| Histidine translase| Histidine tRNA ligase| Histidine tRNA ligase homolog| Histidine tRNA ligase like| Histidine--tRNA ligase| Histidine--tRNA ligase-like| Histidyl tRNA synthetase 2| Histidyl tRNA synthetase 2 mitochondrial| Histidyl tRNA synthetase like| HO3| mitochondrial| Probable histidyl tRNA synthetase mitochondrial| Probable histidyl-tRNA synthetase| SYHM_HUMAN
- 4631412B19Rik|AI593507|HARS related|HARSL|HARSR|HisRS|Histidine translase|Histidine tRNA ligase|Histidine tRNA ligase homolog|Histidine tRNA ligase like|Histidine--tRNA ligase|Histidine--tRNA ligase-like|Histidyl tRNA synthetase 2|Histidyl tRNA synthetase 2 mitochondrial|Histidyl tRNA synthetase like|HO3|mitochondrial|Probable histidyl tRNA synthetase mitochondrial|Probable histidyl-tRNA synthetase|SYHM_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49590
- P49590
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable histidine--tRNA ligase, mitochondrial protein (34-506AA)
- Recombinant Human Probable histidine--tRNA ligase, mitochondrial protein (34-506AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together