Anti-LY96 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: LY96
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-252
- 77742-488
- Antigen Symbol:
- LY96
- Antigen Synonyms:
- ESOP 1|ESOP-1|ESOP1 |LY 96|Ly-96|LY96|LY96_HUMAN|Lymphocyte antigen 96|md 2|MD 2 protein|MD2 protein|Myeloid differentiation protein 2 |Protein MD 2|Protein MD-2|Protein MD2
- ESOP 1|ESOP-1|ESOP1|LY 96|Ly-96|LY96|LY96_HUMAN|Lymphocyte antigen 96|md 2|MD 2 protein|MD2 protein|Myeloid differentiation protein 2|Protein MD 2|Protein MD-2|Protein MD2
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q9Y6Y9
- Q9Y6Y9
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Lymphocyte antigen 96 protein (19-160AA)
- Recombinant Human Lymphocyte antigen 96 protein (19-160AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together