Anti-ENTPD3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ENTPD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77735-944
- 77854-760
- Antigen Symbol:
- ENTPD3
- ENTPD3
- Antigen Synonyms:
- Ecto apyrase|Ecto ATP diphosphohydrolase|CD39L3|Ecto-ATPDase 3|CD39 antigen like 3|CD39 antigen-like 3|OTTHUMP00000162079|NTPDase 3|Ectonucleoside triphosphate diphosphohydrolase 3|NTPDase3|CD39 like 3|FLJ93839|Ecto-ATP diphosphohydrolase 3|Ecto-ATPase 3|HB 6|ENTP3_HUMAN|Ecto-apyrase 3|OTTHUMP00000209227|OTTHUMP00000209228|HB6|ATPDase|ENTPD 3
- ATPDase|CD39 antigen like 3|CD39 antigen-like 3|CD39 like 3|CD39L3|Ecto apyrase|Ecto ATP diphosphohydrolase|Ecto-apyrase 3|Ecto-ATP diphosphohydrolase 3|Ecto-ATPase 3|Ecto-ATPDase 3|Ectonucleoside triphosphate diphosphohydrolase 3|ENTP3_HUMAN|ENTPD 3|FLJ93839|HB 6|HB6|NTPDase 3|NTPDase3|OTTHUMP00000162079|OTTHUMP00000209227|OTTHUMP00000209228
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75355
- O75355
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 3 protein (60-338AA)
- Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 3 protein (60-338AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together