Anti-MAP2K3 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: MAP2K3
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-635
- 77859-362
- Antigen Symbol:
- MAP2K3
- MAP2K3
- Antigen Synonyms:
- AW212142| dual specificity mitogen activated protein kinase kinase 3| Dual specificity mitogen-activated protein kinase kinase 3| MAP kinase kinase 3| MAPK ERK kinase 3| MAPK/ERK kinase 3| MAPKK 3| MAPKK3| MEK 3| MEK3| Mitogen activated protein kinase kinase 3| MKK 3| MKK3| mMKK3b| MP2K3_HUMAN| PRKMK 3| PRKMK3| protein kinase, mitogen-activated, kinase 3| SAPK kinase 2| SAPKK 2| SAPKK2| Stress activated protein kinase kinase 2
- AW212142|dual specificity mitogen activated protein kinase kinase 3|Dual specificity mitogen-activated protein kinase kinase 3|MAP kinase kinase 3|MAPK ERK kinase 3|MAPK/ERK kinase 3|MAPKK 3|MAPKK3|MEK 3|MEK3|Mitogen activated protein kinase kinase 3|MKK 3|MKK3|mMKK3b|MP2K3_HUMAN|PRKMK 3|PRKMK3|protein kinase, mitogen-activated, kinase 3|SAPK kinase 2|SAPKK 2|SAPKK2|Stress activated protein kinase kinase 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P46734
- P46734
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 3 protein (1-170AA)
- Recombinant Human Dual specificity mitogen-activated protein kinase kinase 3 protein (1-170AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together