Anti-GDF11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: GDF11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
- Catalog No:
- 77738-116
- 77855-893
- Antigen Symbol:
- GDF11
- GDF11
- Antigen Synonyms:
- Growth/differentiation factor 11 (GDF-11) (Bone morphogenetic protein 11) (BMP-11), Gdf11, Bmp11
- Growth/differentiation factor 11 (GDF-11) (Bone morphogenetic protein 11) (BMP-11), Gdf11, Bmp11
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Pig
- Pig
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BG54
- Q9BG54
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Pig Growth differentiation factor 11 protein (296-404AA)
- Recombinant Pig Growth differentiation factor 11 protein (296-404AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together