Anti-GSTM3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GSTM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-168
- 77739-167
- Antigen Symbol:
- GSTM3
- GSTM3
- Antigen Synonyms:
- brain GST|brain type mu glutathione S transferase|glutathione S alkyltransferase M3|glutathione S aryltransferase M3|glutathione S transferase M3 (brain)|glutathione S transferase M3|Glutathione S transferase Mu 3|glutathione S transferase, Mu 3|Glutathione S-transferase Mu 3|GST class mu 3|GST class-mu 3|GST5|GSTB|Gstm3|GSTM3-3|GSTM3_HUMAN|GTM3|hGSTM3-3|MGC3310|MGC3704|S (hydroxyalkyl)glutathione lyase M3
- brain GST|brain type mu glutathione S transferase|glutathione S alkyltransferase M3|glutathione S aryltransferase M3|glutathione S transferase M3 (brain)|glutathione S transferase M3|Glutathione S transferase Mu 3|glutathione S transferase, Mu 3|Glutathione S-transferase Mu 3|GST class mu 3|GST class-mu 3|GST5|GSTB|Gstm3|GSTM3-3|GSTM3_HUMAN|GTM3|hGSTM3-3|MGC3310|MGC3704|S (hydroxyalkyl)glutathione lyase M3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21266
- P21266
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Glutathione S-transferase Mu 3 protein (1-225AA)
- Glutathione S-transferase Mu 3 protein (1-225AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together