Anti-NLRP6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NLRP6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-523
- 77744-524
- Antigen Symbol:
- NLRP6
- NLRP6
- Antigen Synonyms:
- Angiotensin II/vasopressin receptor|AVR|CLR11.4|NACHT|leucine rich repeat and PYD containing 6|LRR and PYD containing protein 6|LRR and PYD domains-containing protein 6|NAVR|NAVR/AVR|NLR family|pyrin domain containing 6|NLRP6_HUMAN|Nucleotide binding oligomerization domain|leucine rich repeat and pyrin domain containing 6|PAN3|PYPAF5|PYRIN containing APAF1 like protein 5|PYRIN-containing APAF1-like protein 5
- Angiotensin II/vasopressin receptor|AVR|CLR11.4|NACHT|leucine rich repeat and PYD containing 6|LRR and PYD containing protein 6|LRR and PYD domains-containing protein 6|NAVR|NAVR/AVR|NLR family|pyrin domain containing 6|NLRP6_HUMAN|Nucleotide binding oligomerization domain|leucine rich repeat and pyrin domain containing 6|PAN3|PYPAF5|PYRIN containing APAF1 like protein 5|PYRIN-containing APAF1-like protein 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P59044
- P59044
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NACHT, LRR and PYD domains-containing protein 6 protein (154-306AA)
- NACHT, LRR and PYD domains-containing protein 6 protein (154-306AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together