Order Entry
United States
ContactUsLinkComponent
 
Anti-GSC Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
 
undefined
Anti-GSC Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product

 

  • Catalog No:
  • 77856-499
  • 77739-090
  • Antigen Symbol:
  • GSC
  • GSC
  • Antigen Synonyms:
  • Goosecoid|goosecoid homeobox|GSC_HUMAN|Homeobox protein goosecoid
  • Goosecoid|goosecoid homeobox|GSC_HUMAN|Homeobox protein goosecoid
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P56915
  • P56915
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Homeobox protein goosecoid protein (43-127AA)
  • Recombinant Human Homeobox protein goosecoid protein (43-127AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together