Anti-INTS9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: INTS9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77741-141
- 77858-152
- Antigen Symbol:
- INTS9
- INTS9
- Antigen Synonyms:
- CPSF|CPSF2|CPSF2L|FLJ10871|Int9|INT9_HUMAN|Integrator complex subunit 9|INTS 9|Protein related to CPSF subunits of 74 kDa|RC 74|RC-74|RC74|Related to CPSF subunits 74 kDa
- CPSF|CPSF2|CPSF2L|FLJ10871|Int9|INT9_HUMAN|Integrator complex subunit 9|INTS 9|Protein related to CPSF subunits of 74 kDa|RC 74|RC-74|RC74|Related to CPSF subunits 74 kDa
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NV88
- Q9NV88
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Integrator complex subunit 9 protein (368-658AA)
- Recombinant Human Integrator complex subunit 9 protein (368-658AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together