Anti-PEX11A Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PEX11A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-781
- 77793-782
- Antigen Symbol:
- PEX11A
- PEX11A
- Antigen Name:
- Peroxisomal membrane protein 11A protein (106-219AA)
- Peroxisomal membrane protein 11A protein (106-219AA)
- Antigen Synonyms:
- 28 kDa peroxisomal integral membrane protein|HsPEX11p|MGC119947|MGC138534|Peroxin-11A|Peroxisomal biogenesis factor 11 alpha|Peroxisomal biogenesis factor 11A|Peroxisomal membrane protein 11A|PEX11-alpha|PEX11A|PMP28|Protein PEX11 homolog alpha|PX11A_HUMAN
- 28 kDa peroxisomal integral membrane protein|HsPEX11p|MGC119947|MGC138534|Peroxin-11A|Peroxisomal biogenesis factor 11 alpha|Peroxisomal biogenesis factor 11A|Peroxisomal membrane protein 11A|PEX11-alpha|PEX11A|PMP28|Protein PEX11 homolog alpha|PX11A_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75192
- O75192
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together