Anti-SYT16 Rabbit Polyclonal Antibody
Catalog # 77809-385
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of Synaptotagmin XVI.
- Antigen Symbol:SYT16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q17RD7
- Antigen Synonyms:SYT16|STREP14|SYT14L|SYT14R|Synaptotagmin-16|Chr14Syt|Synaptotagmin 14-like protein|Synaptotagmin XIV-related protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: SYT16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human