Anti-THEMIS2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: THEMIS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-185
- 77829-186
- Antigen Symbol:
- THEMIS2
- THEMIS2
- Antigen Name:
- Protein THEMIS2 protein (62-177AA)
- Protein THEMIS2 protein (62-177AA)
- Antigen Synonyms:
- Basement membrane induced|Chromosome 1 open reading frame 38|ICB 1|ICB1|Induced by contact to basement membrane 1 protein|Protein ICB 1|Protein ICB-1|Protein THEMIS2|THEMIS2|THMS2_HUMAN|Thymocyte expressed molecule involved in selection protein 2|Thymocyte-expressed molecule involved in selection protein 2
- Basement membrane induced|Chromosome 1 open reading frame 38|ICB 1|ICB1|Induced by contact to basement membrane 1 protein|Protein ICB 1|Protein ICB-1|Protein THEMIS2|THEMIS2|THMS2_HUMAN|Thymocyte expressed molecule involved in selection protein 2|Thymocyte-expressed molecule involved in selection protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5TEJ8
- Q5TEJ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together