Anti-TSC22D3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TSC22D3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-019
- 77835-018
- Antigen Symbol:
- TSC22D3
- TSC22D3
- Antigen Name:
- TSC22 domain family protein 3 protein (58-134AA)
- TSC22 domain family protein 3 protein (58-134AA)
- Antigen Synonyms:
- Delta sleep-inducing peptide immunoreactor|DIP|DSIP immunoreactive leucine zipper protein|DSIP-immunoreactive peptide|Dsip1|Dsipi|GILZ|Glucocorticoid-induced leucine zipper protein|hDIP|Protein DIP|T22D3_HUMAN|TSC-22-like protein|TSC-22-related protein|TSC-22R|TSC22 domain family protein 3|TSC22 domain family, member 3|TSC22 related inducible leucine zipper 3|TSC22D3
- Delta sleep-inducing peptide immunoreactor|DIP|DSIP immunoreactive leucine zipper protein|DSIP-immunoreactive peptide|Dsip1|Dsipi|GILZ|Glucocorticoid-induced leucine zipper protein|hDIP|Protein DIP|T22D3_HUMAN|TSC-22-like protein|TSC-22-related protein|TSC-22R|TSC22 domain family protein 3|TSC22 domain family, member 3|TSC22 related inducible leucine zipper 3|TSC22D3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99576
- Q99576
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together