Order Entry
United States
ContactUsLinkComponent
 
Anti-RELN Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
 
undefined
Anti-RELN Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product

 

  • Catalog No:
  • 77797-029
  • 77797-030
  • Antigen Symbol:
  • RELN
  • RELN
  • Antigen Name:
  • Reelin protein (26-254AA)
  • Reelin protein (26-254AA)
  • Antigen Synonyms:
  • LIS2|PRO1598|Reeler|Reelin|RELN|RELN_HUMAN|RL
  • LIS2|PRO1598|Reeler|Reelin|RELN|RELN_HUMAN|RL
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P78509
  • P78509
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together