Anti-PKLR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PKLR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-245
- 77794-244
- Antigen Symbol:
- PKLR
- PKLR
- Antigen Name:
- Pyruvate kinase PKLR protein (1-574AA)
- Pyruvate kinase PKLR protein (1-574AA)
- Antigen Synonyms:
- EC 2.7.1.40|KPYR_HUMAN|L-PK|Pk-1|PK1|PKL|Pklg|Pklr|PKR|PKRL|Pyruvate kinase 1|Pyruvate kinase isozymes R/L|Pyruvate kinase liver and blood cell|Pyruvate kinase liver and RBC|Pyruvate kinase liver and red blood cell|Pyruvate kinase liver type|Pyruvate kinase type L|Pyruvate kinase, red cell type|R type/L type pyruvate kinase|R-PK|R-type/L-type pyruvate kinase|Red cell/liver pyruvate kinase|RPK
- EC 2.7.1.40|KPYR_HUMAN|L-PK|Pk-1|PK1|PKL|Pklg|Pklr|PKR|PKRL|Pyruvate kinase 1|Pyruvate kinase isozymes R/L|Pyruvate kinase liver and blood cell|Pyruvate kinase liver and RBC|Pyruvate kinase liver and red blood cell|Pyruvate kinase liver type|Pyruvate kinase type L|Pyruvate kinase, red cell type|R type/L type pyruvate kinase|R-PK|R-type/L-type pyruvate kinase|Red cell/liver pyruvate kinase|RPK
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30613
- P30613
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together