Anti-PTGES3 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: PTGES3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
- Catalog No:
- 77820-182
- 77820-183
- Antigen Symbol:
- PTGES3
- PTGES3
- Antigen Name:
- Fusion protein of PTGES3
- Fusion protein of PTGES3
- Antigen Synonyms:
- Co chaperone p23|cPGES|Cytosolic prostaglandin E synthase|Cytosolic prostaglandin E2 synthase|Hsp90 co chaperone|Hsp90 co-chaperone|P23|Progesterone receptor complex|Progesterone receptor complex p23|Prostaglandin E synthase 3 (cytosolic)|Prostaglandin E synthase 3|PTGES 3|PTGES3|Sid 3177|TEBP|TEBP_HUMAN|Telomerase binding protein p23|Telomerase-binding protein p23|Unactive progesterone receptor 23 kD
- Co chaperone p23|cPGES|Cytosolic prostaglandin E synthase|Cytosolic prostaglandin E2 synthase|Hsp90 co chaperone|Hsp90 co-chaperone|P23|Progesterone receptor complex|Progesterone receptor complex p23|Prostaglandin E synthase 3 (cytosolic)|Prostaglandin E synthase 3|PTGES 3|PTGES3|Sid 3177|TEBP|TEBP_HUMAN|Telomerase binding protein p23|Telomerase-binding protein p23|Unactive progesterone receptor 23 kD
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15185
- Q15185
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together