Anti-RAB11B Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAB11B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-692
- 77822-693
- Antigen Symbol:
- RAB11B
- RAB11B
- Antigen Name:
- Ras-related protein Rab-11B protein (2-215AA)
- Ras-related protein Rab-11B protein (2-215AA)
- Antigen Synonyms:
- GTP binding protein YPT3|GTP-binding protein ypt3|H-YPT3|RAB 11A|RAB11B|RAB11B member RAS oncogene family|member of RAS oncogene family|Ras related protein Rab 11B|RAS-associated protein RAB11B|Ras-related protein Rab-11B|RB11B_HUMAN|YPT3
- GTP binding protein YPT3|GTP-binding protein ypt3|H-YPT3|RAB 11A|RAB11B|RAB11B member RAS oncogene family|member of RAS oncogene family|Ras related protein Rab 11B|RAS-associated protein RAB11B|Ras-related protein Rab-11B|RB11B_HUMAN|YPT3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15907
- Q15907
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together