Anti-ARAP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-257
- 77834-258
- Antigen Symbol:
- ARAP1
- ARAP1
- Antigen Name:
- Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1 protein (69-344AA)
- Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1 protein (69-344AA)
- Antigen Synonyms:
- 2410002L19Rik| ANK repeat and PH domain-containing protein 1| ARAP 1| Arap1| ARAP1_HUMAN| Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domains containing protein 1| Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domain containing protein 1| Arf-GAP with Rho-GAP domain| CENT D2| Centaurin delta 2| Centaurin delta2| Centaurin-delta-2| CENTD 2| CENTD2| Cnt d2| Cnt-d2| Cntd2| KIAA0782| MGC176140| mKIAA0782
- 2410002L19Rik| ANK repeat and PH domain-containing protein 1| ARAP 1| Arap1| ARAP1_HUMAN| Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domains containing protein 1| Arf GAP Rho GAP ankyrin repeat and pleckstrin homology domain containing protein 1| Arf-GAP with Rho-GAP domain| CENT D2| Centaurin delta 2| Centaurin delta2| Centaurin-delta-2| CENTD 2| CENTD2| Cnt d2| Cnt-d2| Cntd2| KIAA0782| MGC176140| mKIAA0782
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96P48
- Q96P48
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together