Anti-TMLHE Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TMLHE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-170
- 77803-170
- Antigen Symbol:
- TMLHE
- TMLHE
- Antigen Name:
- Trimethyllysine dioxygenase, mitochondrial protein (16-376AA)
- Trimethyllysine dioxygenase, mitochondrial protein (16-376AA)
- Antigen Synonyms:
- AUTSX6| BBOX2| Butyrobetaine (gamma) 2 oxoglutarate dioxygenase (gamma butyrobetaine hydroxylase) 2| Epsilon-trimethyllysine 2-oxoglutarate dioxygenase| Epsilon-trimethyllysine hydroxylase| FLJ10727| TML dioxygenase| TML hydroxylase| TML-alpha-ketoglutarate dioxygenase| TMLD| TMLH| TMLH_HUMAN| TMLHE| TMLHED| Trimethyllysine dioxygenase, mitochondrial| Trimethyllysine hydroxylase epsilon| XAP130
- AUTSX6| BBOX2| Butyrobetaine (gamma) 2 oxoglutarate dioxygenase (gamma butyrobetaine hydroxylase) 2| Epsilon-trimethyllysine 2-oxoglutarate dioxygenase| Epsilon-trimethyllysine hydroxylase| FLJ10727| TML dioxygenase| TML hydroxylase| TML-alpha-ketoglutarate dioxygenase| TMLD| TMLH| TMLH_HUMAN| TMLHE| TMLHED| Trimethyllysine dioxygenase, mitochondrial| Trimethyllysine hydroxylase epsilon| XAP130
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVH6
- Q9NVH6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together