Anti-CA10 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: CA10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77839-826
- 77839-827
- Antigen Symbol:
- CA10
- CA10
- Antigen Name:
- CA10 protein (61-320AA)
- CA10 protein (61-320AA)
- Antigen Synonyms:
- CA 10|CA-RP X|CA10|CAH10_HUMAN|Carbonic anhydrase related protein|Carbonic anhydrase-related protein 10|Carbonic anhydrase-related protein X|CARP X|CARPX|Cerebral protein 15|HUCEP 15
- CA 10|CA-RP X|CA10|CAH10_HUMAN|Carbonic anhydrase related protein|Carbonic anhydrase-related protein 10|Carbonic anhydrase-related protein X|CARP X|CARPX|Cerebral protein 15|HUCEP 15
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NS85
- Q9NS85
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together