Order Entry
United States
ContactUsLinkComponent
Anti-PLEKHF2 Rabbit Recombinant Polyclonal Antibody
Anti-PLEKHF2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-PLEKHF2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77842-092
  • 77842-091
  • Antigen Symbol:
  • PLEKHF2
  • PLEKHF2
  • Antigen Name:
  • Pleckstrin homology domain-containing family F member 2 protein (1-249AA)
  • Pleckstrin homology domain-containing family F member 2 protein (1-249AA)
  • Antigen Synonyms:
  • 110070J07Rik|AA673237|FLJ13187|PH and FYVE domain containing protein 2|PH and FYVE domain-containing protein 2|PH domain containing family F member 2|PH domain-containing family F member 2|Phafin 2|Phafin-2|PHAFIN2|PKHF2_HUMAN|pleckstrin homology domain containing, family F (with FYVE domain) member 2|pleckstrin homology domain containing, family F (with FYVE domain) member2|Pleckstrin homology domain-containing family F member 2|Plekhf2|ZFYVE18|Zinc finger FYVE domain containing protein 18|Zinc finger FYVE domain-containing protein 18
  • 110070J07Rik|AA673237|FLJ13187|PH and FYVE domain containing protein 2|PH and FYVE domain-containing protein 2|PH domain containing family F member 2|PH domain-containing family F member 2|Phafin 2|Phafin-2|PHAFIN2|PKHF2_HUMAN|pleckstrin homology domain containing, family F (with FYVE domain) member 2|pleckstrin homology domain containing, family F (with FYVE domain) member2|Pleckstrin homology domain-containing family F member 2|Plekhf2|ZFYVE18|Zinc finger FYVE domain containing protein 18|Zinc finger FYVE domain-containing protein 18
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9H8W4
  • Q9H8W4
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together