Anti-TCIRG1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TCIRG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77822-901
- 77822-902
- Antigen Symbol:
- TCIRG1
- TCIRG1
- Antigen Name:
- V-type proton ATPase 116 kDa subunit a isoform 3 protein (76-182AA)
- V-type proton ATPase 116 kDa subunit a isoform 3 protein (76-182AA)
- Antigen Synonyms:
- a3| Atp 6i| Atp6i| ATP6N1C| ATP6V0A3| ATPase H+ transporting 116kD| OC 116| OC 116 kDa| OC 116kDa| OC-116 kDa| OC116| OPTB 1| OPTB1| Osteoclastic proton pump 116 kDa subunit| Specific 116 kDa vacuolar proton pump subunit| Stv 1| Stv1| T cell immune regulator 1| T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A| T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A3| T cell immune response cDNA 7| T cell immune response cDNA7 protein| T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a| T-cell immune regulator 1| T-cell immune response cDNA7 protein| TCIRG 1| TCIRG1| TIRC 7| TIRC7| V ATPase 116 kDa| V ATPase 116 kDa isoform a3| V type proton ATPase 116 kDa subunit a| V-ATPase 116 kDa isoform a3| V-type proton ATPase 116 kDa subunit a isoform 3| Vacuolar proton translocating ATPase 116 kDa subunit A| Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3| Vph 1| Vph1| VPP3_HUMAN
- a3| Atp 6i| Atp6i| ATP6N1C| ATP6V0A3| ATPase H+ transporting 116kD| OC 116| OC 116 kDa| OC 116kDa| OC-116 kDa| OC116| OPTB 1| OPTB1| Osteoclastic proton pump 116 kDa subunit| Specific 116 kDa vacuolar proton pump subunit| Stv 1| Stv1| T cell immune regulator 1| T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A| T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A3| T cell immune response cDNA 7| T cell immune response cDNA7 protein| T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a| T-cell immune regulator 1| T-cell immune response cDNA7 protein| TCIRG 1| TCIRG1| TIRC 7| TIRC7| V ATPase 116 kDa| V ATPase 116 kDa isoform a3| V type proton ATPase 116 kDa subunit a| V-ATPase 116 kDa isoform a3| V-type proton ATPase 116 kDa subunit a isoform 3| Vacuolar proton translocating ATPase 116 kDa subunit A| Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3| Vph 1| Vph1| VPP3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13488
- Q13488
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together