Anti-PEX5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PEX5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-811
- 77793-812
- Antigen Symbol:
- PEX5
- PEX5
- Antigen Name:
- Peroxisomal targeting signal 1 receptor protein (149-283AA)
- Peroxisomal targeting signal 1 receptor protein (149-283AA)
- Antigen Synonyms:
- FLJ50634| FLJ50721| FLJ51948| PBD2A| PBD2B| Peroxin 5| Peroxin-5| Peroxisomal biogenesis factor 5| Peroxisomal C terminal targeting signal import receptor| Peroxisomal C-terminal targeting signal import receptor| Peroxisomal targeting signal 1 (SKL type) receptor| Peroxisomal targeting signal 1 receptor| Peroxisomal targeting signal import receptor| Peroxisomal targeting signal receptor 1| Peroxisome receptor 1| pex5| PEX5_HUMAN| PTS1 BP| PTS1 receptor| PTS1-BP| PTS1R| PXR1
- FLJ50634| FLJ50721| FLJ51948| PBD2A| PBD2B| Peroxin 5| Peroxin-5| Peroxisomal biogenesis factor 5| Peroxisomal C terminal targeting signal import receptor| Peroxisomal C-terminal targeting signal import receptor| Peroxisomal targeting signal 1 (SKL type) receptor| Peroxisomal targeting signal 1 receptor| Peroxisomal targeting signal import receptor| Peroxisomal targeting signal receptor 1| Peroxisome receptor 1| pex5| PEX5_HUMAN| PTS1 BP| PTS1 receptor| PTS1-BP| PTS1R| PXR1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50542
- P50542
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together