Anti-TBCB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TBCB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-547
- 77806-546
- Antigen Symbol:
- TBCB
- TBCB
- Antigen Name:
- Tubulin-folding cofactor B protein (1-244AA)
- Tubulin-folding cofactor B protein (1-244AA)
- Antigen Synonyms:
- CG22|CKAPI|Cytoskeleton associated protein 1|Cytoskeleton associated protein CKAPI|Cytoskeleton-associated protein 1|Cytoskeleton-associated protein CKAPI|Tbcb|TBCB_HUMAN|Tubulin folding cofactor B|Tubulin specific chaperone B|Tubulin-folding cofactor B|Tubulin-specific chaperone B
- CG22|CKAPI|Cytoskeleton associated protein 1|Cytoskeleton associated protein CKAPI|Cytoskeleton-associated protein 1|Cytoskeleton-associated protein CKAPI|Tbcb|TBCB_HUMAN|Tubulin folding cofactor B|Tubulin specific chaperone B|Tubulin-folding cofactor B|Tubulin-specific chaperone B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99426
- Q99426
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together