Anti-CTU1 Rabbit Polyclonal Antibody
Catalog # 77810-459
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the C-terminal region of ATPBD3.
- Antigen Symbol:CTU1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q7Z7A3
- Antigen Synonyms:ATP binding domain 3|ATP-binding domain-containing protein 3|Cancer associated protein|Cancer-associated gene protein|ctu1|CTU1_HUMAN|Cytoplasmic tRNA 2-thiolation protein 1|Cytoplasmic tRNA adenylyltransferase 1|Cytosolic thiouridylase subunit 1 homolog|NCS6
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:20000
Type:
Antigen: CTU1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human